National Marker™ Visi-Sign™ Wall Signs

3-D signs project at a 45° angle from the wall to provide 180° visibility

Manufacturer: National Marker Co.Supplier Diversity Partner VS 1 R

 View more versions of this product

Catalog No. 17-984-11A

  Check Availability
Add to Cart

    Specifications

    Height (English) 5.75 in.,5.75 in.
    Quantity Each
    Style Wall Mount
    Color White on Red,White on Red
    Width (English) 8.75 in.,8.75 in.
    Material Acrylic
    For Use With (Application) 3D Mounted signs project 8.9cm from a wall or similar surface at a 45° angle, providing 180° visibility,3D Mounted signs project 8.9cm from a wall or similar surface at a 45° angle, providing 180° visibility
    Message Fire Extinguisher
    Type Safety awareness sign

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit