National Marker Write-On Hazard Warning Labels

Manufacturer: National Marker Co.Supplier Diversity Partner WOL 3

 View more versions of this product

Catalog No. 17-986-90B

  Check Availability
Add to Cart

    Specifications

    Display Text Hazard Warning
    Format Label
    Height (English) 1 in.
    For Use With (Application) The durable adhesive will stick to most flat, clean surfaces, Adhesive Vinyl signs will work for short term outdoor applications and in moderate temperature
    Height (Metric) 2.54cm
    Material Paper
    Product Type Label
    Quantity 500 Roll(s)
    Color Red, Yellow, Blue, White
    Width (English) 3 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit