Thermo Scientific™ National Microvolume Inserts for 8 x 40mm Shell Vials
Thermo Scientific™ National Microvolume Inserts are available in pulled point or precision point design to suit various application needs.
Manufacturer: Thermo Scientific™ C4015536
 View more versions of this product
Catalog No. 03-375-24T
  Check Availability
Add to Cart
    Specifications
    For Use With 8 x 40mm Shell Vials
    Vial Volume (Metric) 275µL
    Type Conical Glass
    Features Pulled Point Tip
    Thermo Scientific™ National Microvolume Inserts are available in pulled point or precision point design to suit various application needs.
    • Polyspring™ inserts feature a polyethylene spring to center the bottom of the insert and to provide a cushion against needle contact
    • Precision point inserts permit maximum sample extraction
    • Use metal spring with inserts that lack the Polyspring
  • Description & Specifications
Related Products
Product Content Feedback