Thermo Scientific National Preassembled Target DP™ Kits
Thermo Scientific™ National Preassembled Target DP Kits include 100 vials with pre-attached caps and septa.
Manufacturer: Thermo Scientific C4000-192W
 View more versions of this product
Catalog No. 03-377-11B
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Type Assembled Vial Kit
    Closure Color Blue
    Septa Red PTFE/White Silicone
    Color Clear glass with ID patch
    Vial Closure Size 9-425mm
    Vial Volume (Metric) 2mL
    Item Description Clear ID; W/PTFE/Silicone Septa
    Description Clear Glass, ID patch; Blue Cap; Red PTFE/White Silicone septum
    Thermo Scientific™ National Preassembled Target DP Kits include 100 vials with pre-attached caps and septa.
    • Packaged in convenient vial trays with clear covers or in polybags
  • Description & Specifications
Related Products
Product Content Feedback