Thermo Scientific National Preassembled Target DP™ Kits
Thermo Scientific™ National Preassembled Target DP Kits include 100 vials with pre-attached caps and septa.
Manufacturer: Thermo Scientific C4000-180W
 View more versions of this product
Catalog No. 03-377-9B
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Septa PTFE/red rubber
    Type Assembled Vial Kit
    Vial Closure Size 9-425mm
    Vial Volume (Metric) 2mL
    Closure Color Blue
    Description PTFE/red rubber septa; Clear glass I-D
    Color Clear glass with ID patch
    Item Description Clear ID; W/PTFE/Red Rubber Septa
    Thermo Scientific™ National Preassembled Target DP Kits include 100 vials with pre-attached caps and septa.
    • Packaged in convenient vial trays with clear covers or in polybags
  • Description & Specifications
Related Products
Product Content Feedback