Thermo Scientific National Preassembled Target DP™ Kits
Thermo Scientific™ National Preassembled Target DP Kits include 100 vials with pre-attached caps and septa.
Manufacturer: Thermo Scientific C4000188WR
 View more versions of this product
Catalog No. 03-375-2
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Septa Red PTFE/White Silicone/Red PTFE Septa
    Color Amber glass with ID patch
    Type Assembled Vial Kit
    Vial Closure Size 9-425mm
    Vial Volume (Metric) 2mL
    Closure Color Red
    Item Description Amber ID; PTFE/Silicone/Red PTFE Septa
    Description Amber Glass I-D; Red
    Thermo Scientific™ National Preassembled Target DP Kits include 100 vials with pre-attached caps and septa.
    • Packaged in convenient vial trays with clear covers or in polybags
  • Description & Specifications
Related Products
Product Content Feedback