Learn More
National Scientific™ BioStor™ Multi-Purpose Specimen Containers
These containers with re-sealable lids can be used to store almost any liquid, powder or solid, making them ideal for laboratory or hospital use.
Supplier: National Scientific™ MPCE T064
Description
Injection molded of polypropylene or linear polyethylene, these containers are virtually indestructible in normal use
- Inert to most lab chemicals including formaldehyde, weak acids, organic solvents and all bases
- Stable, wide-mouth design allows for ease of filling and is less likely to tip over, even when stacked
- Tight-fitting, snap-cap lids keep odors in and minimize chance of leakage
- Contents are visible on contact with translucent plastic walls
- Resistant to boiling and freezing temperatures
Specifications
Specimen Container with Snap-on Lid | |
Tall | |
6 in. |
HDPE | |
64.0 oz. |