Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ National Scientific™ BioStor™ Screw Cap Vials, Conical
Thick-walled construction, with reinforced support collar, allows high speed centrifugation to 17,000xg
Supplier: Thermo Scientific™ BC20NA BP
Description
Multi-purpose design serves a broad spectrum of bio-technology laboratory requirements including the collection, processing, centrifugation, long-term storage, and OEM packaging/shipping of restriction enzymes or other DNA/RNA modifying reagents.
- Circular 3mm syringe port in cap which is easily pierced for radioisotope labeling and fraction collecting
- Inert, LN2-grade polypropylene protects critical sample viability at extreme temperatures ranging from +121C to -195C (LN2 vapor phase)
- Two-turn closure design ensures that tightening of unique screw-cap will uniformly compress “O”-ring against beveled seat, eliminating distortion and maintaining seal integrity
- Vials, caps and ethylene/propylene “O”-rings chemial-resistant and autoclavable with cap loosened
- Molded-in volume lines and etched writing panels on vials and caps will not crack off or dissolve
- External ribs on vials position and lock into serrated holes in metal BioStor Racks, allowing single-handed opening and closing of screw-caps
- Available in natural and ainbow pack
Specifications
Natural | |
2 mL | |
Reinforced support collar, flexible ethylene or propylene O-ring, external rib, circular 3mm syringe port | |
-195°C to +121°C | |
Autoclavable | |
1000/Cs. |
Non-sterile | |
Collection, processing, centrifugation, long term storage and OEM packaging or shipping of restriction enzymes or other DNA or RNA modifying reagents | |
249° to -319°F | |
0.06 oz. | |
Screw Cap |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction