National Scientific BioStor™ Screw Cap Vials, Conical

Thick-walled construction, with reinforced support collar, allows high speed centrifugation to 17,000xg

Manufacturer: National Scientific Supply Company, Inc BC20AS BP

 View more versions of this product

Catalog No. 11-844-17

  Check Availability
Add to Cart

    Specifications

    Color Rainbow
    Capacity (English) 0.06 oz.
    Type Conical Vial
    Closure Type Screw cap
    Sterile No
    Includes Reinforced support collar, flexible ethylene or propylene O-ring, external rib, circular 3mm syringe port
    Capacity (Metric) 2mL
    For Use With (Application) Collection, processing, centrifugation, long term storage and OEM packaging or shipping of restriction enzymes or other DNA or RNA modifying reagents
    Capacity 0.06 oz., 2mL
    Temperature Range (English) 249° to -319°F
    Temperature Range (Metric) 121° to -195°C
    Quantity 1000/Cs.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit