Thermo Scientific™ National Scientific BioStor™ Screw Cap Vials, Conical

Thick-walled construction, with reinforced support collar, allows high speed centrifugation to 17,000xg

Manufacturer: Thermo Scientific™ BC16NA BP

 View more versions of this product

Catalog No. 11-844-8

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Autoclavable Yes
    Color Natural
    Capacity (English) 0.05 oz.
    Type Conical Vial
    Closure Type Screw cap
    Sterile No
    Includes Reinforced support collar, flexible ethylene or propylene O-ring, external rib, circular 3mm syringe port
    Capacity (Metric) 1.6mL
    For Use With (Application) Collection, processing, centrifugation, long term storage and OEM packaging or shipping of restriction enzymes or other DNA or RNA modifying reagents
    Capacity 0.05 oz., 1.6mL
    Quantity 1000/Cs.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit