Learn More
National Scientific™ BioStor™ Screw Cap Vials, Skirted
Thick-walled construction, with reinforced support collar, allows high speed centrifugation to 17,000xg
Manufacturer:  National Scientific™ BS20AS BP
 View more versions of this product
Catalog No. 11-844-19
Add to cart
Description
Description
Multi-purpose design serves a broad spectrum of bio-technology laboratory requirements including the collection, processing, centrifugation, long-term storage, and OEM packaging/shipping of restriction enzymes or other DNA/RNA modifying reagents.
  • Circular 3mm syringe port in cap which is easily pierced for radioisotope labeling and fraction collecting
  • Inert, LN2-grade polypropylene protects critical sample viability at extreme temperatures ranging from +121C to -195C (LN2 vapor phase)
  • Two-turn closure design ensures that tightening of unique screw-cap will uniformly compress “O”-ring against beveled seat, eliminating distortion and maintaining seal integrity
  • Vials, caps and ethylene/propylene “O”-rings chemial-resistant and autoclavable with cap loosened
  • Molded-in volume lines and etched writing panels on vials and caps will not crack off or dissolve
  • External ribs on vials position and lock into serrated holes in metal BioStor Racks, allowing single-handed opening and closing of screw-caps
  • Available in natural and ainbow pack
Specifications
Specifications
Rainbow
2 mL
Reinforced support collar, flexible ethylene or propylene O-ring, external rib, circular 3mm syringe port
-195°C to +121°C
Screw Cap
Nonsterile
Collection, processing, centrifugation, long term storage and OEM packaging or shipping of restriction enzymes or other DNA or RNA modifying reagents
-319°F to +249°F
0.06 oz.
1000/Cs.