Learn More
National Scientific™ Microcentrifuge Tubes
Manufacturer:  National Scientific™ CB1700 GT
 View more versions of this product
Catalog No. 11-842-24
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
RNase, DNase, Endotoxin-free
1.7mL
Polypropylene
Ideal for custom gene libraries, protein work, cDNA synthesis, and nucleic acid amplification
Yes
Blue
17000xG
Yes