Learn More
Thermo Scientific™ National Screw Thread Caps with Bonded Septa Available on GSA/VA Contract for Federal Government customers only.
Effectively eliminate septa push-through with Thermo Scientific™ National Screw Thread Caps with Bonded Septa.
Manufacturer:  Thermo Scientific™ C401069A
 View more versions of this product
Catalog No. 03-342-009
Add to cart
Description
Description
Effectively eliminate septa push-through with Thermo Scientific™ National Screw Thread Caps with Bonded Septa.
  • Septa and polypropylene caps bonded together for an integral system
  • Eliminates septa push-through
  • Available with choice of GPI thread sizes and preslit septa for easier thin-gauge needle penetration
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Black
10-425
Silicone/PTFE
Bonded Red PTFE/White Silicone, 0.005 in.
White
Yes
Polypropylene
Black; Bonded Preslit Red 0.005 in. PTFE/White Silicone Septa; GPI Thread Finish: 10-425
National™
Screw Cap with Septum
PTFE, Silicone