Learn More
Thermo Scientific™ National Screw Thread Caps with Bonded Septa Available on GSA/VA Contract for Federal Government customers only.
Effectively eliminate septa push-through with Thermo Scientific™ National Screw Thread Caps with Bonded Septa.
$52.90 - $61.80
Specifications
Closure Color Black
Septum Bonded Red PTFE/White Silicone, 0.005 in.
View More Specs
Products 3
Catalog Number Mfr. No. Closure Size Septum Price Quantity & Availability  
Catalog Number Mfr. No. Closure Size Septum Price Quantity & Availability  
03-342-006 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
C401068A
10-425 Bonded Red PTFE/White Silicone, 0.005 in. Pack of 100 for $52.90
Add to cart
03-342-009
View Documents
Thermo Scientific™
C401069A
10-425 Bonded Red PTFE/White Silicone, 0.005 in. Pack of 100 for $57.10
Add to cart
03-342-005 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
C401365A
8-425mm Bonded Red PTFE/White Silicone, 0.005 in. Pack of 100 for $61.80
Add to cart
Description
Description
Effectively eliminate septa push-through with Thermo Scientific™ National Screw Thread Caps with Bonded Septa.
  • Septa and polypropylene caps bonded together for an integral system
  • Eliminates septa push-through
  • Available with choice of GPI thread sizes and preslit septa for easier thin-gauge needle penetration
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Black
Bonded Red PTFE/White Silicone, 0.005 in.