Learn More
Thermo Scientific™ National Snap-It™ Closures for 12 x 32 crimp/snap vials
Thermo Scientific™ National Snap-It Closures allow quick, easy application and removal from Target Snap-It vials.
Manufacturer:  Thermo Scientific™ C401156
 View more versions of this product
Catalog No. 03-377-77
  Check Availability
Add to cart
Description
Description
Thermo Scientific™ National Snap-It Closures allow quick, easy application and removal from Target Snap-It vials.
  • Caps eliminate the need for crimping or de-capping tools
  • Easy to apply and easy to remove from vials
  • Available in a variety of colors and septa choices
Specifications
Specifications
Clear
Viton™ Elastomer (Septa)
11mm
C4011-56
National Snap-It™
Clear (Cap)
Viton™ Elastomer
Snap Cap with Septum
2mL