Learn More
Thermo Scientific™ National Snap-It™ Closures for 12 x 32 crimp/snap vials Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ National Snap-It Closures allow quick, easy application and removal from Target Snap-It vials.
Manufacturer:  Thermo Scientific™ C401157
 View more versions of this product
Catalog No. 03-397-20
Add to cart
Description
Description
Thermo Scientific™ National Snap-It Closures allow quick, easy application and removal from Target Snap-It vials.
  • Caps eliminate the need for crimping or de-capping tools
  • Easy to apply and easy to remove from vials
  • Available in a variety of colors and septa choices
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Clear
C4011-57
Integral-Star Slit
Yes
11mm
National Snap-It™
Star-Slit Septum Snap Cap
2 mL