Thermo Scientific™ National Storerooms with Target™ Crimp-Top Vials
Your choice of mini (six-drawer) or full size (nine-drawer) storage units complete with vials and caps.
Manufacturer: Thermo Scientific™ C407521
 View more versions of this product
Catalog No. 03-376-19C
  Check Availability
Add to Cart
    Specifications
    Septa PTFE/RR Seals
    Color Clear ID
    Type NSC Storeroom
    Closure Color Clear
    Closure Type Crimp-Top
    Item Description Storeroom, Clear I-D w/PTFE/RR Seal
    Description NSC Storeroom, Clear I-D Vial
    Thermo Scientific™ National™ Storerooms with Target Crimp-Top Vials feature your choice of mini (six-drawer) or full size (nine-drawer) storage units complete with vials and caps.
    • Mini Storeroom has 6 drawers; each drawer contains 100 packs of vials or septa
    • Full size storeroom has 9 deep drawers with a capacity of 2000 vials and seals
    • Stackable cabinet constructed of heavy-gauge steel
    • With metal shelves and injection-molded drawers
    • Storeroom can be restocked as product is consumed
    Includes:Prestocked with 3000 each of: clear vials and preassembled clear crimp seals with FEP/red rubber septa
  • Description & Specifications
Related Products
Product Content Feedback