Thermo Scientific™ National Target™ Assembled Vial, Cap and Septum Kits
Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
Manufacturer: Thermo Scientific™ C401017BLK
 View more versions of this product
Catalog No. 03-376-432
  Check Availability
Add to Cart
    Specifications
    Septa Red PTFE/White Silicone
    Color Clear
    Type Assembled Vial Kit
    Closure Color Light Blue
    Item Description Clear; Black cap; PTFE/Silicone
    Description Clear Glass; Black cap; PTFE/Silicone septum
    Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
    • Assembled kits include 100 vials with pre-attached caps and septa
    • Packaged in convenient vial trays with clear covers or in polybags
    • Vial trays are stackable for easy storage
    • Vial trays hold vials upright for easy filling and maintaining sample order
  • Description & Specifications
Related Products
Product Content Feedback