Thermo Scientific™ National Target™ Assembled Vial, Cap and Septum Kits Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
Manufacturer: Thermo Scientific™ C4010057
 View more versions of this product
Catalog No. 03-375-44A
 N/A
    N/A
Part Not Found 
    Specifications
    Septa PTFE/Silicone, Shimadzu
    Closure Color White
    Description Clear Glass; White Cap; PTFE/Silicone septum, Shimadzu
    Color Clear
    Type Assembled Vial Kit
    Vial Closure Size 10-425mm
    Vial Volume (Metric) 2mL
    Item Description Clear; PTFE/Silicone septum, Shimadzu
    Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
    • Assembled kits include 100 vials with pre-attached caps and septa
    • Packaged in convenient vial trays with clear covers or in polybags
    • Vial trays are stackable for easy storage
    • Vial trays hold vials upright for easy filling and maintaining sample order
  • Description & Specifications
Related Products
Product Content Feedback