Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific National Target DP™ Robovial™ Caps with Pre-inserted Septa

Minimize contamination from handling with convenient Thermo Scientific™ National Target DP Robovial Caps with Pre-Inserted Septa.

Manufacturer:  Thermo Scientific C4000-62R

 View more versions of this product

Catalog No. 03-377-506

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Specifications

Description

Minimize contamination from handling with convenient Thermo Scientific™ National Target DP Robovial Caps with Pre-Inserted Septa.

  • Bonded caps feature a PTFE/Silicone septum electrolytically bonded into the cap, reducing septa push-through when used with blunt tip needles on high-speed autosamplers
  • Pre-assembled caps and septa are convenient to minimize contamination from handling
  • Fully compatible with all Target DP vials
  • Easy-on, easy-off convenience with just half-a-turn
  • Storage cap has a solid top and pre-inserted PTFE/red rubber liner
Description & Specifications

Specifications

Target DP vials
Red
Bonded Natural PTFE/Clear Silicone
Red
Red
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit