Learn More
Thermo Scientific National Target DP™ Screw Caps with Septa
Thermo Scientific™ National Target DP Screw Caps with Septa are color coded to separate assays, standards or samples.
Manufacturer:  Thermo Scientific C400051A
 View more versions of this product
Catalog No. 03-396-96
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
Thermo Scientific™ National Target DP Screw Caps with Septa are color coded to separate assays, standards or samples.
  • Preassembled with septa
Description & Specifications
Specifications
Open Top Screw Cap
C4000-51A
Ivory PTFE/Red Rubber
Ivory PTFE/Red Rubber Septa; Black
Black
9-425mm