Thermo Scientific National Target DP™ Screw Caps with Septa
Thermo Scientific™ National Target DP Screw Caps with Septa are color coded to separate assays, standards or samples.
Manufacturer: Thermo Scientific C400057G
 View more versions of this product
Catalog No. 03-375-77B
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Description Pre-slit Ivory PTFE/Red Rubber Septa; Green
    Closure Color Green
    Pre-slit Yes
    Septum Ivory PTFE/Red Rubber
    Vial Closure Size 9-425mm
    Type Open Top Screw Cap
    Item Description C4000-57G
    Thermo Scientific™ National Target DP Screw Caps with Septa are color coded to separate assays, standards or samples.
    • Preassembled with septa
  • Description & Specifications
Related Products
Product Content Feedback