Thermo Scientific National Target DP™ Screw Thread Caps
Thermo Scientific™ National Target DP Screw Thread Caps with open top are designed for use with Target DP septa.
Manufacturer: Thermo Scientific C4000-98B
 View more versions of this product
Catalog No. 03-377-4A
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Description Blue
    Closure Color Blue
    Closure Material Polypropylene
    Septum No Septum
    Vial Closure Size 9-425mm
    Type Open Top Screw Cap
    For Use With Target DP septa
    Item Description Blue
    Thermo Scientific™ National Target DP Screw Thread Caps with open top are designed for use with Target DP septa.
    • Polypropylene, open-top caps for use with Target DP septa
  • Description & Specifications
Related Products
Product Content Feedback