Learn More
Thermo Scientific™ National Target DP™ Screw Thread Caps Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ National Target DP Screw Thread Caps with open top are designed for use with Target DP septa.
Manufacturer:  Thermo Scientific™ C500098G
 View more versions of this product
Catalog No. 03-377-4B
  Check Availability
Add to cart
Specifications
Description
Thermo Scientific™ National Target DP Screw Thread Caps with open top are designed for use with Target DP septa.
  • Polypropylene, open-top caps for use with Target DP septa
Description & Specifications
Specifications
Target DP septa
Green
Green
No Septum
Open Top Screw Cap
Green
Polypropylene