Thermo Scientific National Target DP™ Vial Kits
Thermo Scientific™ National Target DP Vial Kits include vials and caps with preassembled septa in a convenient benchtop storage tray.
Manufacturer: Thermo Scientific C4000-91AW
 View more versions of this product
Catalog No. 03-376-422
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Septa Red PTFE/White Silicone
    Closure Color Black
    Item Description C4000-91AW
    Color Amber ID
    Type Assembled Vial Kit
    Description Amber Glass I-D; Red PTFE/White Silicone septum
    Thermo Scientific™ National Target DP Vial Kits include vials and caps with preassembled septa in a convenient benchtop storage tray.
    • Convenience Kits with 2mL, 12 x 32mm vials
    • I-D vials feature write-on patch with graduations at 0.5, 1.0 and 1.5mL
    • Blue caps with your choice of vial and preassembled seals with septa
    • Packaged in two-compartment benchtop storage tray
    • All items remain dust-free
    • Caps with pre-inserted septa are not attached to vials (packed in separate polybag)
  • Description & Specifications
Related Products
Product Content Feedback