Learn More
Thermo Scientific National Target DP™ Vial Kits
Thermo Scientific™ National Target DP Vial Kits include vials and caps with preassembled septa in a convenient benchtop storage tray.
Manufacturer:  Thermo Scientific C4000-92W
 View more versions of this product
Catalog No. 03-377-8B
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
Thermo Scientific™ National Target DP Vial Kits include vials and caps with preassembled septa in a convenient benchtop storage tray.
  • Convenience Kits with 2mL, 12 x 32mm vials
  • I-D vials feature write-on patch with graduations at 0.5, 1.0 and 1.5mL
  • Blue caps with your choice of vial and preassembled seals with septa
  • Packaged in two-compartment benchtop storage tray
  • All items remain dust-free
  • Caps with pre-inserted septa are not attached to vials (packed in separate polybag)
Description & Specifications
Specifications
Assembled Vial Kit
2mL
C4000-92W
Red PTFE/White Silicone
9-425mm
Blue
Clear Glass with I-D; Red PTFE/White Silicone septum
Clear ID