Learn More
Thermo Scientific™ National Target DP™ Vial Kits
Thermo Scientific™ National Target DP Vial Kits include vials and caps with preassembled septa in a convenient benchtop storage tray.
Products 1
Catalog Number Mfr. No. Type Closure Color Septum Color Price Quantity & Availability  
Catalog Number Mfr. No. Type Closure Color Septum Color Price Quantity & Availability  
03-377-7B
View Documents
Thermo Scientific™
C400086W
Blue Red PTFE/White Silicone/Red PTFE Clear This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Thermo Scientific™ National Target DP Vial Kits include vials and caps with preassembled septa in a convenient benchtop storage tray.
  • Convenience Kits with 2mL, 12 x 32mm vials
  • I-D vials feature write-on patch with graduations at 0.5, 1.0 and 1.5mL
  • Blue caps with your choice of vial and preassembled seals with septa
  • Packaged in two-compartment benchtop storage tray
  • All items remain dust-free
  • Caps with pre-inserted septa are not attached to vials (packed in separate polybag)