Thermo Scientific™ National Target™ Snap-It™ Kits
Thermo Scientific™ National Target Snap-It Convenience Kits provide vials and Snap-it caps with prefitted septa in storage trays.
Manufacturer: Thermo Scientific™ C401178P
 View more versions of this product
Catalog No. 03-377-613
  Check Availability
Add to Cart
    Specifications
    Septa PTFE/Silicone
    Material Clear Glass
    Pre-slit Y-cut
    Color Clear
    Type Assembled Vial Kit
    Vial Volume (Metric) 2mL
    Closure Color Pink
    Thermo Scientific™ National Target Snap-It Convenience Kits provide vials and Snap-it caps with prefitted septa in storage trays.
    • 2mL,12 x 32mm vials
    • Choice of vial and preassembled seals with septa
    • Packaged in two compartment benchtop storage trays
    • Include matched quantities of vials and Snap-It caps with prefitted septa
    • All items remain dust-free
  • Description & Specifications
Related Products
Product Content Feedback