Learn More
Thermo Scientific™ National Target™ Snap-It™ Kits
Thermo Scientific™ National Target Snap-It Convenience Kits provide vials and Snap-it caps with prefitted septa in storage trays.
$29.60 - $91.00
Specifications
Volume (Metric) 2 mL
View More Specs
Products 6
Catalog Number Mfr. No. Closure Color Color Pre-slit Septum Price Quantity & Availability  
Catalog Number Mfr. No. Closure Color Color Pre-slit Septum Price Quantity & Availability  
03-377-84
View Documents
Thermo Scientific™
C401172W
Clear Clear PTFE/Red Rubber Pack of 100 for $67.90
Add to cart
03-377-88
View Documents
Thermo Scientific™
C401174
Clear Clear Yes PTFE/Silicone, Pre-slit Pack of 100 for $91.00
Add to cart
03-377-611
View Documents
Thermo Scientific™
C401172P
Pink Clear PTFE/Red Rubber Pack of 100 for $29.60
Add to cart
03-377-86
View Documents
Thermo Scientific™
C401173AW
Clear Amber PTFE/Silicone This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
03-377-89
View Documents
Thermo Scientific™
C401174AW
Clear Amber Yes PTFE/Silicone, Pre-slit This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
03-377-613
View Documents
Thermo Scientific™
C401178P
Pink Clear Y-cut PTFE/Silicone This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Thermo Scientific™ National Target Snap-It Convenience Kits provide vials and Snap-it caps with prefitted septa in storage trays.
  • 2mL,12 x 32mm vials
  • Choice of vial and preassembled seals with septa
  • Packaged in two compartment benchtop storage trays
  • Include matched quantities of vials and Snap-It caps with prefitted septa
  • All items remain dust-free
Specifications
Specifications
2 mL