Thermo Scientific™ National Target™ Snap-It™ Storerooms
Thermo Scientific™ National Target Snap-It Storerooms provide convenient storage for vials and caps.
Manufacturer: Thermo Scientific™ C40116551
 View more versions of this product
Catalog No. 03-339-350
  Check Availability
Add to Cart
    Specifications
    Septa PTFE/RR
    Type Mini Vial Storeroom
    Closure Material PTFE/RR
    Closure Type Snap-It
    Item Description C4011-6551
    Description NSC Mini Storeroom, Clear Snap-It Vial, PTFE/RR Cap, 500 pieces
    Thermo Scientific™ National Target Snap-It Storerooms provide convenient storage for vials and caps.
    • Mini Storeroom has 6 drawers, each drawer will hold one 100/pack of vials or 500/pack caps
    • Full-size Storeroom has 9 deep drawers with a capacity of 2000 vials and caps
    • Storeroom can be restocked as product is consumed
  • Description & Specifications
Related Products
Product Content Feedback