Thermo Scientific National Unassembled Certified™ Vial Kits
Thermo Scientific™ National Unassembled Certified Vial Kits provide extra assurance that time after time, the product upholds rigorous manufacturing standards.
Manufacturer: Thermo Scientific CERT400071LV
 View more versions of this product
Catalog No. 03-377-340
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Type Unassembled Vial Kit
    Septa PTFE/Silica
    Pre-slit Pre-silt
    Color Clear ID
    Vial Closure Size 9-425mm
    Vial Volume (Metric) 350µL
    Closure Material Bonded Slit PTFE/Silicone
    Closure Type Screw Thread
    Item Description DP 350µL Clear I-D Fused Insert; Slit
    Description Certified Target DP 350µL Clear Glass I-D Fused Insert Vial, 9mm Screw Thread
    Thermo Scientific™ National Unassembled Certified Vial Kits provide extra assurance that time after time, the product upholds rigorous manufacturing standards.
    • Fully lot tested for HPLC and GC analysis
    • Certificate of Conformance in every pack
    • Bonded caps to prevent septa push-through
    • Clear Type 1, 33 expansion borosilicate glass or amber Type 2, 51 expansion borosilicate glass
    • Available with or without ID patch
    • Fixed micro vials or inserts for limited-volume sampling
  • Description & Specifications
Related Products
Product Content Feedback