Learn More
Thermo Scientific™ National Unassembled Certified™ Vial Kits
Thermo Scientific™ National Unassembled Certified Vial Kits provide extra assurance that time after time, the product upholds rigorous manufacturing standards.
$83.00 - $113.80
Products 2
Catalog Number Mfr. No. Volume (Metric) Closure Material Closure Type Color Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Closure Material Closure Type Color Price Quantity & Availability  
03-377-354
View Documents
Thermo Scientific™
CERT401586
4 mL Bonded PTFE/Silicone Screw Thread Amber
Pack of 100 for $113.80
Add to cart
03-377-351
View Documents
Thermo Scientific™
CERT401599
1 mL Polyethylene SepCap Amber
Pack of 200 for $83.00
Add to cart
Description
Description
Thermo Scientific™ National Unassembled Certified Vial Kits provide extra assurance that time after time, the product upholds rigorous manufacturing standards.
  • Fully lot tested for HPLC and GC analysis
  • Certificate of Conformance in every pack
  • Bonded caps to prevent septa push-through
  • Clear Type 1, 33 expansion borosilicate glass or amber Type 2, 51 expansion borosilicate glass
  • Available with or without ID patch
  • Fixed micro vials or inserts for limited-volume sampling