Learn More
SP Industries National Undercounter Glassware Washers: NLW-88 Basic
Clean glassware is essential. A National Undercounter Glassware Washer is made for use in university, pharmaceutical, bioresearch, chemical research, clinical, and industrial environments.
Manufacturer:  SP Industries NLW88
 View more versions of this product
Catalog No. 15-365-41
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
24 in.
Ideal for university, pharmaceutical, bioresearch, chemical research, clinical and industrial environments
115V
19 in.
19 x 21.5 x 25 in. (48.2 x 54.6 x 63.5cm)
25 in.
63.5cm
Type 304 Stainless Steel
Undercounter Washer
44.9 gal.
6
165 lb.
21.5 in.
54.6cm
ISO 9001:2000, ETL ETLc
60.96cm
60Hz
15A
48.2cm
115V 60Hz
34.25 to 36.5 in.
86.9 to 92.7cm
18 to 60psi for tap water
29° to 77°C
170L
9 to 15L (Tap Water)
75kg
24 in.
60.96cm