Thermo Scientific™ National Vial Crushers
Reduce waste disposal costs by reducing the total volume of waste with the Thermo Scientific™ National Vial Crusher.
Manufacturer: Thermo Scientific™ C4040PVCM
 View more versions of this product
Catalog No. 03-376-245
  Check Availability
Add to Cart
    Specifications
    Description Pneumatic Benchtop Vial Crusher with Stainless-steel Mobile Cart
    Type Vial Accessories
    Item Description Pneumatic benchtop vial crusher with stainless-steel mobile cart
    Reduce waste disposal costs by reducing the total volume of waste with the Thermo Scientific™ National Vial Crusher.
    • Breaks and separates liquid from all 1mL to 100mL vials and ampules
    • Capable of processing approximately 5000 2mL vials per hour
    • Safe use in a fume hood or connect to a vacuum for limited exposure to flammable or toxic vapors
    • Environmentally responsible waste handling—crushed glass is automatically separated from solvent for separate disposal
  • Description & Specifications
Related Products
Product Content Feedback