Learn More
Thermo Scientific™ National Vial Crushers Available on GSA/VA Contract for Federal Government customers only.
Reduce waste disposal costs by reducing the total volume of waste with the Thermo Scientific™ National Vial Crusher.
Manufacturer:  Thermo Scientific™ C4040VCMC
Catalog No. 03-376-247
Add to cart
Description
Description
Reduce waste disposal costs by reducing the total volume of waste with the Thermo Scientific™ National Vial Crusher.
  • Breaks and separates liquid from all 1mL to 100mL vials and ampules
  • Capable of processing approximately 5000 2mL vials per hour
  • Safe use in a fume hood or connect to a vacuum for limited exposure to flammable or toxic vapors
  • Environmentally responsible waste handling—crushed glass is automatically separated from solvent for separate disposal
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.