Learn More
Thermo Scientific™ WebSeal™ Microplate Sealing System
The Thermo Scientific™ National WebSeal Microplate Sealing System provides superior resealability after multiple injections.
Supplier: Thermo Scientific™ M9601RF2P
Description
The Thermo Scientific™ National WebSeal Microplate Sealing System provides superior resealability after multiple injections.
When productivity really counts and cross-contamination simply won't be tolerated, choose the WebSeal System for efficiency and security. WebSeal system products are used in high-throughput screening, combinatorial chemistry, life science and HPLC. This system is a comprehensive range of 96-well microtiter plates with glass inserts and silicone/PTFE sealing mats.
- Manufactured from pure silicone with PTFE coating on underside
- Excellent chemical compatibility
- Dry heat autoclavable
- Eliminates cross-contamination well to well
- Available pre-slit for easy penetration and reduced vacuum formation
Specifications
Pink | |
96 | |
Microplate Sealing Components | |
Round |
PTFE-coated Silicone | |
10 | |
Webseal | |
96-well Plate |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.