Learn More
Thermo Scientific™ WebSeal™ Microplate Sealing System
The Thermo Scientific™ National WebSeal Microplate Sealing System provides superior resealability after multiple injections.
$162.10 - $260.50
Specifications
Material Silicone with PTFE Coating
View More Specs
Products 2
Catalog Number Mfr. No. No. of Wells Well Shape Material Color Price Quantity & Availability  
Catalog Number Mfr. No. No. of Wells Well Shape Material Color Price Quantity & Availability  
03-397-81
View Documents
Thermo Scientific™
M9601RF2P
96 Round, Flat Base Silicone with PTFE Coating Pink
Pack of 10 for $260.50
Add to cart
03-397-84
View Documents
Thermo Scientific™
M9601SQ2Y
96 Square Silicone with PTFE Coating Yellow
Pack of 10 for $162.10
Add to cart
Description
Description
The Thermo Scientific™ National WebSeal Microplate Sealing System provides superior resealability after multiple injections.
When productivity really counts and cross-contamination simply won't be tolerated, choose the WebSeal System for efficiency and security. WebSeal system products are used in high-throughput screening, combinatorial chemistry, life science and HPLC. This system is a comprehensive range of 96-well microtiter plates with glass inserts and silicone/PTFE sealing mats.
  • Manufactured from pure silicone with PTFE coating on underside
  • Excellent chemical compatibility
  • Dry heat autoclavable
  • Eliminates cross-contamination well to well
  • Available pre-slit for easy penetration and reduced vacuum formation
Specifications
Specifications
Silicone with PTFE Coating