Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Native Green Fluorescent Protein Rabbit anti-Cnidarian, Polyclonal, Bii Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

$139.52 - $514.84

Specifications

Antigen Native Green Fluorescent Protein
Concentration 1 mg/mL
Applications ELISA, Immunoblot, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0654 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
BIINGFPAB4100
0.1 mL
Each for $514.84
Add to cart
 
50-210-0655 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
BIINGFPAB425
250 μg
Each for $139.52
Add to cart
 
Description

Description

Native Green Fluorescent Protein Polyclonal antibody specifically detects Native Green Fluorescent Protein in Cnidarian samples. It is validated for ELISA, Immunoblot, Immunocytochemistry, Western Blot
Specifications

Specifications

Native Green Fluorescent Protein
ELISA, Immunoblot, Immunocytochemistry, Western Blot
Unconjugated
Rabbit
Cnidarian
GFP; GFP tag; GFP2; green fluorescence; green fluorescent; Turbo eGFP
IgG
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
1 mg/mL
Polyclonal
Liquid
RUO
10mM tris with 0.02% sodium azide; pH
native Green Fluorescent Protein (Immunoaffinity purified)
Polyclonal
Affinity chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit