Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Natriuretic Peptide Receptor C Mouse anti-Human, Mouse, Rat, HRP, Clone: OTI4C3, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272881H
Description
Natriuretic Peptide Receptor C Monoclonal specifically detects Natriuretic Peptide Receptor C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Natriuretic Peptide Receptor C | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
NPR3 | |
Full length recombinant protein of human Natriuretic Peptide Receptor C (NP_000899) produced in HEK293T cells. | |
0.1 mL | |
4883.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4C3 | |
Western Blot, Immunohistochemistry | |
ANP-C, ANPR-C, ANPRCFLJ14054, Atrial natriuretic peptide clearance receptor, atrial natriuretic peptide receptor 3, Atrial natriuretic peptide receptor type C, C5orf23, chromosome 5 open reading frame 23, GUCY2B, natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptidereceptor C), NPRC, NPR-C | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title