Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix NATtrol™ BK Virus External Run Controls

Click to view available options
Concentration:
10000 copies/mL
1000000 copies/mL
Description
- Contains 6 x 1.0mL vials of BK Virus NATtrol™ at concentrations of 10,000 to 1,000,000 copies/mL
- Supplied in purified protein matrix identical to the composition of true clinical specimen
Contain intact virus and should be run in a manner identical to that used for clinical specimens.
Validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Specifications
Specifications
Certifications/Compliance | ISO, 13485 |
Concentration | 1000000 copies/mL |
Content And Storage | 2°C to 8°C |
Form | Solution |
Media Type | Vial |
Product Type | NATtrol External Run Control |
Formulation | Formulated with purified, intact virus particles |
For Use With (Application) | Determination of presence of Human BK virus DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel |
Quantity | 6 x 1 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction