Learn More
Zeptometrix NATtrol™ CT/NG/TV Controls
Designed to evaluate the performance of nucleic acid tests for determination of the presence of C. trachomatis, N. gonorrhoeae, and T. vaginalis DNA.
Supplier:  Zeptometrix NATCTNGTVNEG
 View more versions of this product
Catalog No. 22-158-434
Add to Cart
Description
Description
NATtrol™ CT/NG/TV Positive Control is formulated with purified, intact microorganisms. The microorganisms have been chemically modified to render them non-infectious and refrigerator stable. NATtrol™ CT/NG/TV Negative Control is formulated in the absence of any microorganisms. Each control pack contains 6 x 1.0 mL of NATtrol™ CT/NG/TV Positive Control or NATtrol™ CT/NG/TV Negative Control. These controls are provided in a proprietary matrix.
INTENDED USE:
  • NATtrol™ CT/NG/TV Positive Control and NATtrol™ CT/NG/TV Negative Control are designed to evaluate the performance of nucleic acid tests for determination of the presence of C. trachomatis, N. gonorrhoeae, and T. vaginalis DNA.
  • NATtrol™ CT/NG/TV Positive Control and NATtrol™ CT/NG/TV Negative Control enable laboratories to monitor test variation, lot-to-lot test kit performance, operator variation, and can provide assistance in identifying random or systemic error.
Specifications
Specifications
Negative Controls
Evaluate performance of molecular tests, verification of assays, training of laboratory personnel and to monitor assay-kit lot performance
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
RUO only.