Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix NATtrol™ Epstein-Barr Virus, B95-8 Strain
Designed to evaluate the performance of nucleic acid tests for determination of the presence of EBV DNA
Supplier: ZeptoMetrix NATEBV0002
Description
- Formulated with purified, intact virus particles which are chemically modified to render them non-infectious and refrigerator stable
- Supplied in a purified protein matrix that mimics the composition of a real clinical specimen
- Contains 6 x 1.0mL vials
- Available in different concentrations: 100, 500, 1000, 10000, 50000, 100000 copies/mL
NATEBV contains intact virus and should be run in a manner identical to that used for clinical specimens
Validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Specifications
ISO, 13485 | |
2°C to 8°C | |
Vial | |
Formulated with purified, intact virus particles | |
1 mL |
500 copies/mL | |
Solution | |
NATtrol Virus Control | |
Determination of presence of EBV DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction