Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix NATtrol™ Human Hepatitis B
Ideal control for amplification assays
Specifications
Certifications/Compliance | ISO, 13485 |
---|---|
Content And Storage | 2°C to 8°C |
Form | Solution |
Media Type | Vial |
Product Type | NATtrol Virus Stock |
Catalog Number | Mfr. No. | Concentration | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Concentration | Price | Quantity & Availability | |||||
22-157-785
|
ZeptoMetrix
NATHBV0003 |
50000IU/mL |
Each for $672.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-783
|
ZeptoMetrix
NATHBV0001 |
400IU/mL |
Each for $64.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-784
|
ZeptoMetrix
NATHBV0002 |
5000IU/mL |
Each for $214.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-786
|
ZeptoMetrix
NATHBV0006 |
10000IU/mL |
Each for $393.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Formulated from highly purified infectious viruses, from cell or plasma of infected individuals
- After purification virus is chemically and enzymatically modified to alter surface proteins
- Provide completely intact viral particle that is unable to bind, penetrate or infect any host cell
Human Source Material used in the manufacture of this product has been tested and found to be negative for HBsAg, antibodies to HIV-1/2 and HCV. Handle as if capable of transmitting infectious agents; use Universal Precautions when working with this product.
Used as a control/calibrators for PCR, TMA, Branched Chain DNA and NAT assays
Specifications
ISO, 13485 | |
Solution | |
NATtrol Virus Stock | |
Determination of presence of HBV DNA, use as controls and calibrators for PCR, TMA, Branched Chain DNA and other NAT assays |
2°C to 8°C | |
Vial | |
Formulated with purified, intact virus particles | |
1 mL |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title