Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix NATtrol™ Chlamydia trachomatis LGVII 434 External Run Controls
Formulated with purified, intact chemically modified virus particles to render them non-infectious and refrigerator stable
$582.00
Specifications
Certifications/Compliance | ISO, 13485 |
---|---|
Content And Storage | 2°C to 8°C |
Form | Solution |
Media Type | Vial |
Product Type | NATtrol External Run Control |
Catalog Number | Mfr. No. | Concentration | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Concentration | Price | Quantity & Availability | |||||
22-156-721
|
ZeptoMetrix
NATCT(434)ERCM |
50000 copies/mL |
Each for $582.00
|
|
|||||
22-157-773
|
ZeptoMetrix
NATCT(434)ERCL |
5000 copies/mL |
Each for $481.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Contains 6 x 1.0mL vials of C. trachomatis NATtrol™ at concentrations of 5000 to 50,000 copies/mL
- Supplied in purified protein matrix identical to the composition of true clinical specimen
Contain intact organisms and should be run in a manner identical to that used for clinical specimens.
Validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Specifications
ISO, 13485 | |
Solution | |
NATtrol External Run Control | |
Determination of presence of C. trachomatis DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel |
2°C to 8°C | |
Vial | |
Formulated with purified, intact bacterial cells | |
6 x 1 mL |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title