Fisherbrand™ Natural HDPE Wide-Mouth Oblong Bottles with White Polypropylene PE Foam Lined Cap
Ideal for sampling solids and powders. Fisherbrand™ Natural HDPE Wide-Mouth Oblong Bottles with White Polypropylene PE Foam Lined Cap offer a space saving design for the workbench.
 View more versions of this product
Catalog No. 02-911-954A
  Check Availability
Add to Cart
Includes: Polypropylene PE Foam lined cap
    Specifications
    Capacity (Metric) 160mL
    Capacity (English) 5.5 oz.
    Product Type Oblong Bottle
    Closure Color White
    Closure Material Polypropylene
    Closure Type Screw
    Color Natural
    Material HDPE
    Liner Material PE Foam
    Mouth Wide
    Neck Finish 38-400
    Quantity 48/Cs.
    Shape Oblong
    • HDPE provides moisture barrier and are impact resistant
    • Ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping
    • Polypropylene closures with foamed polyethylene liners offer good chemical resistance including acids, alcohols, alkalis, aqueous products, cosmetics, household oils and solvents
  • Description & Specifications
Related Products
Product Content Feedback