Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Natural HDPE Wide Mouth Oblong Bottles with White Polypropylene PE Foam Cap

Ideal for sampling solids and powders. Fisherbrand™ Natural HDPE Wide-Mouth Oblong Bottles with White Polypropylene PE Foam Lined Cap offer a space saving design for the workbench.

 View more versions of this product

Catalog No. 02-911-957A


Add to cart

Includes: White Polypropylene PE foam lined caps

Description

Description

Natural HDPE Wide Mouth Oblong Bottles offer a space saving design for the workbench. These bottles feature a wide mouth opening, ideal for sampling solids and powders. HDPE provides an excellent moisture barrier and are impact resistant. They are ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping.

  • HDPE provides moisture barrier and are impact resistant
  • Ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping
  • Polypropylene closures with foamed polyethylene liners offer good chemical resistance including acids, alcohols, alkalis, aqueous products, cosmetics, household oils and solvents
Specifications

Specifications

250mL
Polypropylene
HDPE, HDPE
Bottle, Oblong, Oblong Bottle
PE Foam
43-400
Screw
8.5 oz.
Natural, Natural
48/Cs.
White
Wide
Oblong
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit