Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Natural HDPE Wide-Mouth Oblong Bottles with White Polypropylene PE Foam Lined Cap

Ideal for sampling solids and powders. Fisherbrand™ Natural HDPE Wide-Mouth Oblong Bottles with White Polypropylene PE Foam Lined Cap offer a space saving design for the workbench.

 View more versions of this product

Catalog No. 02-911-949A


  Check Availability
Add to cart

Includes: Polypropylene PE Foam lined cap

Specifications

Description

  • HDPE provides moisture barrier and are impact resistant
  • Ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping
  • Polypropylene closures with foamed polyethylene liners offer good chemical resistance including acids, alcohols, alkalis, aqueous products, cosmetics, household oils and solvents
Description & Specifications

Specifications

1000mL
Polypropylene
HDPE
Bottle, Oblong
PE Foam
53-400
Screw
32 oz.
Natural
12/Cs.
White
Wide
Oblong
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit