Fisherbrand™ Natural HDPE Wide-Mouth Round Bottles without Cap
Ideal for general lab use with solids and powders. Fisherbrand™ Natural HDPE Wide Mouth Round Bottles without Cap features wide mouth that facilitates easy mixing, handling, and transfer of contents.
 View more versions of this product
Catalog No. 02-911-874A
  Check Availability
Add to Cart
    Specifications
    Product Type Wide Mouth Round Bottle
    Material HDPE
    Mouth Wide
    Neck Finish 89-400
    Capacity (Metric) 3785mL
    Capacity (English) 128 oz.
    Color Natural
    Quantity 4/Cs.
    Shape Round
    • HDPE (high-density polyethylene) bottles provides moisture barrier, are translucent and impact resistant
    • For high-speed filling, demanding pharmaceutical specifications and rough handling of interstate shipping
  • Description & Specifications
Related Products
Product Content Feedback