Fisherbrand™ Natural HDPE Wide-Mouth Round Bottles with White Polypropylene PE Foam Lined Caps
Suggested for use with water based chemicals and some food products. Fisherbrand™ Natural HDPE Wide-Mouth Round Bottles with White Polypropylene PE Foam Lined Cap feature a wide opening making them ideal for solids.
 View more versions of this product
Catalog No. 02-911-990A
  Check Availability
Add to Cart
Includes: Polypropylene PE Foam lined cap
    Specifications
    Capacity (Metric) 2000mL
    Capacity (English) 68 oz.
    Product Type Wide Mouth Round Bottle
    Closure Color White
    Closure Material Polypropylene
    Closure Type Screw
    Color Natural
    Material HDPE
    Liner Material PE Foam
    Mouth Wide
    Neck Finish 89-400
    Quantity 50/Cs.
    Shape Round
    • Translucent bottles allow limited viewing of contents
    • Provides moisture barrier and are impact resistant
  • Description & Specifications
Related Products
Product Content Feedback