Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Natural HDPE Wide-Mouth Round Bottles with White Polypropylene PE Foam Lined Caps

Suggested for use with water based chemicals and some food products. Fisherbrand™ Natural HDPE Wide-Mouth Round Bottles with White Polypropylene PE Foam Lined Cap feature a wide opening making them ideal for solids.

 View more versions of this product

Catalog No. 02-912-032A


  Check Availability
Add to cart

Includes: Polypropylene PE Foam lined cap

Specifications

Description

  • Translucent bottles allow limited viewing of contents
  • Provides moisture barrier and are impact resistant
Description & Specifications

Specifications

120mL
Polypropylene
HDPE
Bottle, Wide Mouth Round
PE Foam
38-400
Screw
4 oz.
Natural
48/Cs.
White
Wide
Round
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit